Merck
  • Apolipoprotein A-I increases insulin secretion and production from pancreatic β-cells via a G-protein-cAMP-PKA-FoxO1-dependent mechanism.
Apolipoprotein A-I increases insulin secretion and production from pancreatic β-cells via a G-protein-cAMP-PKA-FoxO1-dependent mechanism.
Arteriosclerosis, thrombosis, and vascular biology (2014-08-26)
Blake J Cochran, Radjesh J Bisoendial, Liming Hou, Elias N Glaros, Jérémie Rossy, Shane R Thomas, Philip J Barter, Kerry-Anne Rye
ABSTRACT
Therapeutic interventions that increase plasma levels of high-density lipoproteins and apolipoprotein A-I (apoA-I) A-I, the major high-density lipoprotein apolipoprotein, improve glycemic control in people with type 2 diabetes mellitus. High-density lipoproteins and apoA-I also enhance insulin synthesis and secretion in isolated pancreatic islets and clonal β-cell lines. This study identifies the signaling pathways that mediate these effects. Incubation with apoA-I increased cAMP accumulation in Ins-1E cells in a concentration-dependent manner. The increase in cAMP levels was inhibited by preincubating the cells with the cell-permeable, transmembrane adenylate cyclase inhibitor, 2'5' dideoxyadenosine, but not with KH7, which inhibits soluble adenylyl cyclases. Incubation of Ins-1E cells with apoA-I resulted in colocalization of ATP-binding cassette transporter A1 with the Gαs subunit of a heterotrimeric G-protein and a Gαs subunit-dependent increase in insulin secretion. Incubation of Ins-1E cells with apoA-I also increased protein kinase A phosphorylation and reduced the nuclear localization of forkhead box protein O1 (FoxO1). Preincubation of Ins-1E cells with the protein kinase A-specific inhibitors, H89 and PKI amide, prevented apoA-I from increasing insulin secretion and mediating the nuclear exclusion of FoxO1. Transfection of Ins-1E cells with a mutated FoxO1 that is restricted to the nucleus confirmed the requirement for FoxO1 nuclear exclusion by blocking insulin secretion in apoA-I-treated Ins-1E cells. ApoA-I also increased Irs1, Irs2, Ins1, Ins2, and Pdx1 mRNA levels. ApoA-I increases insulin synthesis and secretion via a heterotrimeric G-protein-cAMP-protein kinase A-FoxO1-dependent mechanism that involves transmembrane adenylyl cyclases and increased transcription of key insulin response and β-cell survival genes.
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder