- Firefly luciferase: the structure is known, but the mystery remains.
Firefly luciferase: the structure is known, but the mystery remains.
Structure (London, England : 1993) (1996-03-15)
T O Baldwin
PMID8805542
ABSTRACT
The structure of firefly luciferase reveals a new protein fold which may be representative of a growing family of homologous enzymes.