Merck
  • Anhydrous proton transport system based on zwitterionic liquid and HTFSI.
Anhydrous proton transport system based on zwitterionic liquid and HTFSI.
Chemical communications (Cambridge, England) (2004-08-13)
Masahiro Yoshizawa, Hiroyuki Ohno
ABSTRACT
Novel binary ionic liquids based on zwitterionic liquid and HTFSI for anhydrous proton transport were prepared and showed ionic conductivity of about 10(-2) S cm(-1) at 150 degrees C and a zwitterionic liquid content of 60 mol%.
Sigma-Aldrich
1-Butylimidazole, 98%