Merck
  • Effect of unilateral ureteral obstruction and anti-angiotensin II treatment on renal tubule and interstitial cell apoptosis in rats.
Effect of unilateral ureteral obstruction and anti-angiotensin II treatment on renal tubule and interstitial cell apoptosis in rats.
Croatian medical journal (2008-10-18)
Nikola Radović, Snjezan Cuzić, Mladen Knotek
ABSTRACT
To investigate the effects of angiotensin-converting enzyme inhibitor (cilazapril) and angiotensin II type I receptor antagonist (losartan) on tubular and interstitial cell apoptosis and caspase-3 activity in rats with obstructive nephropathy after unilateral ureteral obstruction. Rats with unilateral obstructive nephropathy and sham-operated rats were treated with cilazapril, losartan, or the vehicle (water). Tubular and interstitial cell apoptosis was detected morphologically on hematoxylin and eosin-stained renal specimens and by the terminal deoxynucleotidyl transferase-mediated nick end-labeling. Caspase-3 activity in whole-kidney tissue homogenates was measured colorimetrically. After unilateral ureter ligation, there was a significant increase in the number of apoptotic tubular and interstitial cells in the obstructed kidney (P=0.049 and P=0.036, respectively, vs sham-operated rats, 10 days after ligation). In rats with unilateral obstructive nephropathy, neither cilazapril nor losartan had an effect on tubular cell apoptosis. However, cilazapril caused a significant increase in the number of renal apoptotic interstitial cells (P=0.019). Caspase-3 activity was not significantly different in rats with unilateral obstructive nephropathy than in sham-operated rats. Rats with unilateral obstructive nephropathy had increased apoptosis of tubular and interstitial cells in comparison with sham-operated rats. Neither cilazapril nor losartan had an effect on tubular cell apoptosis, and cilazapril even increased interstitial cell apoptosis.
Product Description
Cilazapril, European Pharmacopoeia (EP) Reference Standard
Cilazapril impurity D, European Pharmacopoeia (EP) Reference Standard