Merck
  • Molecular cloning and expression analysis of a delta 6-fatty acid desaturase gene from Rhizopus stolonifer strain YF6 which can accumulate high levels of gamma-linolenic acid.
Molecular cloning and expression analysis of a delta 6-fatty acid desaturase gene from Rhizopus stolonifer strain YF6 which can accumulate high levels of gamma-linolenic acid.
Journal of microbiology (Seoul, Korea) (2011-03-04)
Xia Wan, Yinbo Zhang, Ping Wang, Mulan Jiang
ABSTRACT
The delta 6-desaturase gene was cloned from Rhizopus stolonifer, which could accumulate up to 49% of gamma-linolenic acid (GLA, C18:3 Δ(6,9,12)) to the total fatty acids. The cloned DNA contains a 1,380 bp open reading frame encoding a protein of 460 amino acids, which showed high similarity to those of fungal delta 6-desaturases with three conserved histidine-rich motifs and HPGG motif. Notably, this deduced sequence had a shorter C-terminus. Results demonstrated that the cDNA sequence exhibited delta 6-desaturase activity by accumulation of about 22.4 % of GLA to the total fatty acids in the recombinant Pichia pastoris strain GS115.