Merck
  • Complexes of Strong Bidentate Lewis Acids Derived from 2,7-Bis(1,1-dimethylethyl)fluorene-1,8-diol.
Complexes of Strong Bidentate Lewis Acids Derived from 2,7-Bis(1,1-dimethylethyl)fluorene-1,8-diol.
Inorganic chemistry (2001-10-24)
Okba Saied, Michel Simard, James D. Wuest
ABSTRACT
Treatment of 2,7-bis(1,1-dimethylethyl)fluorene-1,8-diol (3) with 2 equiv of TiCl(4) converts the two OH groups into OTiCl(3) groups and thereby yields bis(trichlorotitanium phenoxide) 7, a structurally well-defined reagent that holds two sites of strong Lewis acidity in close proximity. Bidentate Lewis acid 7 forms a crystalline 1:2 complex with 4,4'-dimethylbenzophenone. An X-ray crystallographic study revealed that each atom of titanium binds only one molecule of ketone to form an unusual pentacoordinate adduct with an approximately trigonal bipyramidal geometry. Treatment of fluorenediol 3 with 2 equiv of Al(CH(2)CH(3))(3) does not yield the expected bis(diethylaluminum phenoxide) 8, but rather its dimer 9. Formation of diverse reagents 7 and 9 from the same precursor demonstrates that the strategy of converting organic compounds with suitably oriented hydroxyl groups into the corresponding metal alkoxides is a particularly versatile and effective way to make strong, structurally well-defined multidentate Lewis acids.
Sigma-Aldrich
4,4′-Dimethylbenzophenone, 99%