Merck
  • Beneficial effects of vildagliptin combined with miglitol on glucose tolerance and islet morphology in diet-controlled db/db mice.
Beneficial effects of vildagliptin combined with miglitol on glucose tolerance and islet morphology in diet-controlled db/db mice.
Biochemical and biophysical research communications (2013-10-10)
Keita Ishibashi, Akemi Hara, Yoshio Fujitani, Toyoyoshi Uchida, Koji Komiya, Motoyuki Tamaki, Hiroko Abe, Takeshi Ogihara, Akio Kanazawa, Ryuzo Kawamori, Hirotaka Watada
ABSTRACT
Dipeptidyl peptidase-4 (DPP-4) inhibitors improve glycemic control in patients with type 2 diabetes primarily by increasing plasma active glucagon-like peptide-1 (GLP-1) levels. While various combination therapies based on DPP-4 inhibitors have been proposed for treatment of type 2 diabetes, the effects of combination therapy of DPP-4 inhibitors and alpha-glucosidase inhibitors on β-cell function are less characterized. We evaluated the effects of long-term treatment with vildagliptin, a DPP-4 inhibitor, on metabolic parameters and β-cell function, in combination with miglitol, an alpha-glucosidase inhibitor, in diet-controlled db/db mice. In this study, 6-week-old male db/db mice were provided with standard chow twice a day for 6 weeks. Meal tolerance tests and glucose tolerance tests showed that the combination therapy of vildagliptin with miglitol, but not each alone, suppressed postprandial glycemic excursion, enhanced postprandial active GLP-1 levels and prevented deterioration of glucose tolerance in the db/db mice. The combination treatment did not alter β-cell mass, but resulted in preserved expression of glucose transporter 2, Zinc transporter 8 and MafA and reduced the number of α cells. These results suggest that the combination of vildagliptin and miglitol prevents the development of overt diabetes in diet-controlled pre-diabetic db/db mice by normalizing postprandial glucose and incretin response, and by preserving β-cell structure and the expression of factors essential for β-cell function.
Sigma-Aldrich
1-Deoxynojirimycin hydrochloride
Sigma-Aldrich
Adamantane, ≥99%
Supelco
1-Deoxynojirimycin, analytical standard
Sigma-Aldrich
Miglitol