Merck
Stiripentol in refractory status epilepticus.
Epilepsia (2013-09-06)
Denise K Grosenbaugh, David D Mott
ABSTRACT
Benzodiazepines (BZDs), which enhance γ-aminobutyric acid (GABAA ) receptor-mediated inhibition, are the first-line therapy for treatment of status epilepticus (SE). However, pharmacoresistance to BZDs develops rapidly after SE initiation. This is due to an activity-dependent internalization of BZD-sensitive GABAA receptors during SE. Stiripentol (STP) is a positive allosteric modulator of GABAA receptors with a unique subunit selectivity profile. We report that in a rodent model of SE, STP terminates behavioral seizures and remains effective in established SE when seizures have become BZD resistant. The anticonvulsant effects of STP are age dependent, with greater potency in juvenile animals. Whole cell recordings from dentate granule cells in hippocampal slices reveal that STP potentiates GABAergic inhibitory postsynaptic currents (IPSCs) and tonic GABAergic currents by acting at a site on the GABAA receptor that is separate from the benzodiazepine binding site. This potentiation persists in established SE, whereas potentiation of GABAergic inhibition by BZDs is lost. STP potentiates IPSCs in juvenile animals with greater potency than in adult animals. We suggest that STP, either alone or as add-on therapy, may prove useful in treating established and BZD-resistant status epilepticus. Furthermore, STP may be particularly effective in terminating SE in children when SE is most prevalent.
Sigma-Aldrich
Stiripentol, ≥98% (HPLC)