- Cloning and characterization of KLHL5, a novel human gene encoding a kelch-related protein with a BTB domain.
Cloning and characterization of KLHL5, a novel human gene encoding a kelch-related protein with a BTB domain.
Biochemical genetics (2001-10-10)
S Wang, Z Zhou, K Ying, R Tang, Y Huang, C Wu, Y Xie, Y Mao
PMID11590829
ABSTRACT
Most kelch family proteins contain two conserved domains, the BTB domain and the kelch repeat domain. Here we describe the cloning and characterization of a novel human KLHL5 gene. The 3488 bp cDNA encodes a kelch family protein homologous to the Drosophila kelch protein. It also contains the two conserved domains. Northern blot analysis reveals a single transcript. It is abundantly expressed in ovary, adrenal gland, and thyroid and less abundantly expressed in trachea, prostate, testis, lymph node, and spinal cord. KLHL5 was mapped to 4p13-4p15.1.