Merck
  • Synthesis of diazaphenoxathiin nucleosides from 3-deazauridine and their chemical properties.
Synthesis of diazaphenoxathiin nucleosides from 3-deazauridine and their chemical properties.
Nucleic acids symposium series (1983-01-01)
M Kaneko, M Kimura, Y Murofushi, M J Robins
ABSTRACT
New method for a synthesis of diazaphenoxathiin skeleton from 3-deazauracil derivatives is reported. It became possible to convert 3-deazauridine to 3-deazacytidine via an excellent intermediate "diazaphenoxathiin sulfoxide derivative".