Merck
  • One-pot, two-step synthesis of imidazo[1,2-a]benzimidazoles via a multicomponent [4 + 1] cycloaddition reaction.
One-pot, two-step synthesis of imidazo[1,2-a]benzimidazoles via a multicomponent [4 + 1] cycloaddition reaction.
ACS combinatorial science (2013-09-11)
Ya-Shan Hsiao, Bharat D Narhe, Ying-Sheng Chang, Chung-Ming Sun
ABSTRACT
A one-pot, two-step synthesis of imidazo[1,2-a]benzimidazoles has been achieved by a three-component reaction of 2-aminobenzimidazoles with an aromatic aldehyde and an isocyanide. The reaction involving condensation of 2-aminobenzimidazole with an aldehyde is run under microwave activation to generate an imine intermediate under basic conditions which then undergoes [4 + 1] cycloaddition with an isocyanide.
Sigma-Aldrich
2-Aminobenzimidazole, 97%