- Determination of galactose and mannose residues in natural galactomannans using a fast and efficient high-performance liquid chromatography/UV detection.
Determination of galactose and mannose residues in natural galactomannans using a fast and efficient high-performance liquid chromatography/UV detection.
Journal of chromatography. A (2008-01-08)
N Tapie, C Malhiac, N Hucher, M Grisel
PMID18177885
ABSTRACT
The present work describes the validation of an easy, fast and efficient precolumn derivatization method for the quantification of oligosides, mannose and galactose obtained by degradation of galactomannans. This work combines an acid hydrolysis and an enzymatic degradation of natural galactomannans with the quantification of released residues by reversed-phase HPLC-UV, the most usual HPLC system in laboratories. In case of enzymatic degradation, mannotetraose has been detected and quantified for the first time, and an application to the evaluation of the galactosyl distribution in galactomannans is proposed. After an acidic hydrolysis, this method also allowed to obtain the mannose/galactose (M/G) ratio.
MATERIALS