Merck
  • Inhibition of growth and patulin synthesis in Penicillium expansum by potassium sorbate and sodium propionate in culture.
Inhibition of growth and patulin synthesis in Penicillium expansum by potassium sorbate and sodium propionate in culture.
Applied and environmental microbiology (1984-11-01)
J E Lennox, L J McElroy
ABSTRACT
Potassium sorbate and sodium propionate brought about a marked inhibition in the growth of Penicillium expansum and a proportionally greater inhibition in the synthesis of patulin by the mold. At inhibitor concentrations used commercially in bakery products, propionate inhibited growth less efficiently than sorbate did but was a more effective inhibitor of patulin synthesis.
Sigma-Aldrich
Sodium propionate, meets analytical specification of NF, 99-100.5% (ex dried subst.)