Merck
  • Regulation of priority carcinogens and reproductive or developmental toxicants.
Regulation of priority carcinogens and reproductive or developmental toxicants.
American journal of industrial medicine (1992-01-01)
K Hooper, J LaDou, J S Rosenbaum, S A Book
ABSTRACT
In California, 370 carcinogens and 112 reproductive/developmental toxicants have been identified as a result of the State's Safe Drinking Water and Toxic Enforcement Act of 1986. They include pesticides, solvents, metals, industrial intermediates, environmental mixtures, and reactive agents. Occupational, environmental, and consumer product exposures that involve these agents are regulated under the Act. At levels of concern, businesses must provide warnings for and limit discharges of those chemicals. The lists of chemicals were compiled following systematic review of published data, including technical reports from the U.S. Public Health Service--National Toxicology Program (NTP), and evaluation of recommendations from authoritative bodies such as the International Agency for Research on Cancer (IARC) and the U.S. Environmental Protection Agency (USEPA). Given the large number of chemicals that are carcinogens or reproductive/developmental toxicants, regulatory concerns should focus on those that have high potential for human exposure, e.g., widely distributed or easily absorbed solvents, metals, environmental mixtures, or reactive agents. In this paper, we present a list of 33 potential priority carcinogens and reproductive/developmental toxicants, including alcoholic beverages, asbestos, benzene, chlorinated solvents, formaldehyde, glycol ethers, lead, tobacco smoke, and toluene.
Sigma-Aldrich
3-Chloro-2-methyl-1-propene, technical grade, 90%
Pricing and availability is not currently available.