Merck
  • Synthesis of 14 C-radiolabeled oligonucleotides with a novel phosphoramidite reagent.
Synthesis of 14 C-radiolabeled oligonucleotides with a novel phosphoramidite reagent.
Bioorganic & medicinal chemistry letters (1999-01-01)
A Guzaev, B Boyode, G Balow, K L Tivel, M Manoharan
ABSTRACT
A novel solid-phase synthesis of 5'-radiolabeled oligonucleotides is described. The labeling reaction is carried out by the phosphoramidite method with the aid of [4,6-di-14C]-5'-dimethoxytritylthymidine building block 1. The feasibility of the method is demonstrated by preparation of 3'-phosphorylated dodecathymidylate phosphorothioate containing radiolabeled nucleoside at the 5'-terminus.
Sigma-Aldrich
5′-O-(4,4′-Dimethoxytrityl)thymidine, 98%