- Charge transport along the lambda-DNA double helix.
Charge transport along the lambda-DNA double helix.
Physical review letters (2000-09-06)
P Tran, B Alavi, G Gruner
PMID10970555
ABSTRACT
We have measured the conductivity sigma along the lambda phage DNA (lambda-DNA) double helix at microwave frequencies using lyophilized DNA in and also without a buffer. The conductivity is strongly temperature dependent around room temperature with a crossover to a weakly temperature dependent conductivity at low temperatures. Removal of the water mantle around the double helix leads to reduced conductivity.
Sigma-Aldrich
Lambda Phage DNA, Methylated from Escherichia coli host strain W3110, lyophilized powder