Merck
  • Tyrosine derivatization and preparative purification of the sialyl and asialy-N-linked oligosaccharides from porcine fibrinogen.
Tyrosine derivatization and preparative purification of the sialyl and asialy-N-linked oligosaccharides from porcine fibrinogen.
Archives of biochemistry and biophysics (1994-07-01)
M L Da Silva, T Tamura, T McBroom, K G Rice
ABSTRACT
The N-linked oligosaccharides from porcine fibrinogen were purified following their release from glycopeptides using N-glycosidase F. In separate experiments, both sialyl and asialyl oligosaccharides were prepared from 5 g of fibrinogen. The reducing oligosaccharides were reacted with ammonium bicarbonate to form/oligosaccharide-glycosylamines and then derivatized with tert-butoxycarbonyl-L-tyrosine N-hydroxysuccinimidyl ester. Tyrosinamide--oligosaccharides were purified first by gel filtration chromatography and then by reverse-phase HPLC and the products were characterized by proton NMR and fast atom bombardment-MS. Porcine fibrinogen was found to have predominantly a single asialyl biantennary oligosaccharide containing a fucose linked alpha 1-6 to GlcNAc 1. The oligosaccharide possesses two sialylation patterns with a major form (70%) having a single N-acetyl neuraminic acid (NeuAc) residue linked alpha 2-6 to galactose on only one antenna and a minor form (30%) possessing two NeuAc residues linked alpha 2-6 to both terminal galactose residues. In addition to developing an isolation procedure and establishing the structures of porcine fibrinogen oligosaccharides, this study improves on the tyrosine derivatization technique as a general approach to isolate structurally diverse N-linked oligosaccharides from glycoproteins.
Sigma-Aldrich
Boc-Tyr-OH, 98%