Merck
  • Improved assay of reaction products to quantitate catechol-O-methyltransferase activity by high-performance liquid chromatography with electrochemical detection.
Improved assay of reaction products to quantitate catechol-O-methyltransferase activity by high-performance liquid chromatography with electrochemical detection.
Journal of chromatography. B, Biomedical applications (1995-01-06)
I Reenilä, P Tuomainen, P T Männistö
ABSTRACT
We applied coulometric detection (three electrochemical electrodes in series) to quantitate vanillic acid and isovanillic acid using reversed-phase HPLC. The formation of these reaction products from dihydroxybenzoic acid was used as a precise and reproducible measure of catechol-O-methyltransferase (COMT) activity in striatal homogenates and recombinant membrane-bound COMT protein. This detection system has a higher sensitivity (0.5 pmol per injection) than a single-cell amperometric detection. As in a previous method, the deproteinized supernatants of the COMT assay could be injected directly onto the HPLC system allowing the handling of a large number of samples in one day.