- Tranexamic acid derivatives with enhanced absorption.
Tranexamic acid derivatives with enhanced absorption.
Journal of medicinal chemistry (1986-04-01)
C M Svahn, F Merenyi, L Karlson, L Widlund, M Grälls
PMID3959024
ABSTRACT
Derivatives of the antifibrinolytic drug tranexamic acid [trans-4-(aminomethyl)cyclohexanecarboxylic acid] containing one or two tranexamic acid moieties were synthesized. Most of the derivatives have good stability in acidic and neutral solutions but are easily hydrolyzed in plasma. By measuring the amount of tranexamic acid excreted in the urine after an oral dose, relative absorptions of a number of derivatives in the rat were estimated. Most of the derivatives showed greater absorption than tranexamic acid itself. 1-[(Ethoxycarbonyl)oxy]ethyl trans-4-(amino-methyl)cyclohexanecarboxylate hydrochloride was chosen for studies in man.