- Mephentermine dependence with psychosis. A case report.
Mephentermine dependence with psychosis. A case report.
The British journal of psychiatry : the journal of mental science (1988-01-01)
G J Uday, U G Josh, S M Bhat
PMID3167321
ABSTRACT
Dependence on mephentermine, a widely used sympathomimetic pressor agent, is so far unreported, although misuse of inhalers is recognised. A case of mephentermine dependence associated with chronic psychosis is reported here. Psychosis initially surfaced with chronic dexamphetamine abuse, but was sustained for 3 years by mephentermine. After a period of remission lasting for 4 years, the patient again developed psychosis on restarting abuse of mephentermine, which lasted for 5 years.