Merck
  • Prostaglandin dependence of membrane order changes during myogenesis in vitro.
Prostaglandin dependence of membrane order changes during myogenesis in vitro.
Biochimica et biophysica acta (1988-03-03)
M T Santini, P L Indovina, R E Hausman
ABSTRACT
Myogenic differentiation in vitro involves at least three events at the cell surface: binding of prostaglandin to cells, cell-cell adhesion, and fusion of the myoblast membranes into syncytia. Previous work has suggested that binding of prostaglandin is causal to the change in cell-cell adhesion and that both are accompanied by a characteristic reorganization of the myoblast membrane detected as a transient increase in membrane order by electron paramagnetic resonance. We show here that this membrane order change, which reaches a maximum at 38 h of development in vitro, was the last membrane order change before bilayer fusion which begins several hours later. This membrane order change, which accompanies the change in cell-cell adhesion, was dependent on the availability of prostaglandin. In myoblasts maintained in indomethacin, where further differentiation is known to be blocked at the prostaglandin binding step, the membrane order change did not occur. However, if myoblasts are provided with exogenous prostaglandin, the membrane order change occurred and differentiation proceeded. The results indicate that the basis of the membrane order change was the reorganization of myoblast membranes to allow increased adhesion and prepare the membrane for bilayer fusion. They also demonstrate that, like the increase in myoblast adhesion, the membrane order change was dependent on prostaglandin being available to bind to its receptor.