Merck
  • MODIFICATIONS OF CULTURE MEDIA USED IN THE ISOLATION AND DIFFERENTIATION OF TYPHOID, DYSENTERY, AND ALLIED BACILLI.
MODIFICATIONS OF CULTURE MEDIA USED IN THE ISOLATION AND DIFFERENTIATION OF TYPHOID, DYSENTERY, AND ALLIED BACILLI.
The Journal of experimental medicine (1918-09-01)
I J Kligler
ABSTRACT
The three media as modified-Endo medium, neutral red-brilliant green, and the lead acetate Russell double sugar-have proved in our hands the best combination for the isolation and rapid differentiation of the various organisms belonging to the typhoid-dysentery group of bacilli.
Product Description
Millipore
Kligler Agar, suitable for microbiology