Merck
  • Morphine to codeine concentration ratio in blood and urine as a marker of illicit heroin use in forensic autopsy samples.
Morphine to codeine concentration ratio in blood and urine as a marker of illicit heroin use in forensic autopsy samples.
Forensic science international (2011-12-06)
Svetlana V Konstantinova, Per T Normann, Marianne Arnestad, Ritva Karinen, Asbjørg S Christophersen, Jørg Mørland
ABSTRACT
A morphine to codeine ratio greater than unity (M/C>1) has been suggested as an indicator of heroin use in living individuals. The aim of this study was to examine the morphine to codeine ratio in a large population (N=2438) of forensically examined autopsy cases positive for 6-monoacetylmorphine (6-MAM) and/or morphine in blood and/or urine. Blood and urine concentrations of 6-MAM, morphine and codeine were examined using GC-MS and LC-MS/MS methods. In 6-MAM positive samples, the M/C ratio was greater than unity in 98% (N=917) of the blood samples and 96% (N=665) of the urine samples. Stratification of 6-MAM negative cases by M/C above or below unity revealed similarities in morphine and codeine concentrations in cases where M/C>1 and 6-MAM positive cases. Median blood and urine morphine concentrations were 8-10 times greater than codeine for both groups. Similarly to 6-MAM positive cases, 25-44 year-old men prevailed in the M/C>1 group. In comparison to cases where M/C ≤ 1, the M/C ratio was a hundred times higher in both 6-MAM positive and M/C>1 cases. The range of morphine concentration between the lowest and the highest quintile of codeine in M/C>1 cases was similar to that in 6-MAM positive cases. This range was much higher than for M/C ≤ 1 cases. Moreover, linear regression analyses, adjusted for age and gender, revealed a strong positive association between morphine and codeine in 6-MAM positive and M/C>1 cases. The M/C ratio appeared to be a good marker of heroin use in post-mortem cases. Both blood and urine M/C>1 can be used to separate heroin users from other cases positive for morphine and codeine.