Merck
Fragrance material review on α-isobutylphenethyl alcohol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-11-01)
J Scognamiglio, L Jones, C S Letizia, A M Api
ABSTRACT
A toxicologic and dermatologic review of α-isobutylphenethyl alcohol when used as a fragrance ingredient is presented. α-Isobutylphenethyl alcohol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a secondary alcohol. The AAAs are a structurally diverse class of fragrance ingredients that includes primary, secondary, and tertiary alkyl alcohols covalently bonded to an aryl (Ar) group, which may be either a substituted or unsubstituted benzene ring. The common structural element for the AAA fragrance ingredients is an alcohol group -C-(R1)(R2)OH and generically the AAA fragrances can be represented as an Ar-C-(R1)(R2)OH or Ar-Alkyl-C-(R1)(R2)OH group. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. Available data for α-isobutylphenethyl alcohol were evaluated then summarized and includes physical properties, skin sensitization, and repeated dose data. A safety assessment of the entire Aryl Alkyl Alcohols will be published simultaneously with this document; please refer to Belsito et al. (2012) for an overall assessment of the safe use of this material and all Aryl Alkyl Alcohols in fragrances.
Sigma-Aldrich
α-Isobutylphenethyl alcohol, 98%