Merck
  • The prenylation inhibitor manumycin A reduces the viability of Anaplasma phagocytophilum.
The prenylation inhibitor manumycin A reduces the viability of Anaplasma phagocytophilum.
Journal of medical microbiology (2011-02-26)
Qingming Xiong, Yasuko Rikihisa
ABSTRACT
Anaplasma phagocytophilum is an obligately intracellular bacterium and is the causative agent of human granulocytic anaplasmosis (HGA), an emerging and major tick-borne disease in the USA and other parts of the world. This study showed that the prenylation inhibitor manumycin A effectively blocked A. phagocytophilum infection in host cells (HL-60 or RF/6A cells). A. phagocytophilum infection activated extracellular signal-regulated kinase (ERK) mitogen-activated protein kinase in host cells, and manumycin A treatment reduced ERK activation in A. phagocytophilum-infected host cells. As ERK activation is required for A. phagocytophilum infection, we examined whether manumycin A inhibited the bacteria directly or through host ERK signalling. Treatment of A. phagocytophilum alone with manumycin A significantly reduced the bacterial infectivity of host cells and bacterial viability in the absence of host cells, whereas pre-treatment of host cells did not inhibit bacterial infection in host cells. The inhibitory effect of manumycin A on A. phagocytophilum infection in host cells was achieved even at a concentration 100 times lower than that required for effective inhibition of mammalian cell signalling. These results suggested that manumycin A directly inactivates the bacterium, resulting in reduced infection and ERK1/2 activation. Thus, the manumycin group of drugs may have a therapeutic potential for HGA.
Product Description
Millipore
Phosphatase Inhibitor Cocktail Set II, A cocktail of five phosphatase inhibitors for the inhibition of acid and alkaline phosphatases as well as protein tyrosine phosphatases (PTPs). Suitable for use with cell lysates and tissue extracts.
Sigma-Aldrich
Manumycin A from Streptomyces parvulus
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.