- Degradation of the metal-cyano complex tetracyanonickelate(II) by cyanide-utilizing bacterial isolates.
Degradation of the metal-cyano complex tetracyanonickelate(II) by cyanide-utilizing bacterial isolates.
Ten bacterial isolates capable of growth on tetracyanonickelate(II) [K2[Ni(CN)4]] (TCN) as the sole nitrogen source were isolated from soil, freshwater, and sewage sludge enrichments. Seven of the 10 were identified as pseudomonads, while the remaining 3 were classified as Klebsiella species. A detailed investigation of one isolate, Pseudomonas putida BCN3, revealed a rapid growth rate on TCN (generation time, 2 h), with substrate removal and growth occurring in parallel. In addition to TCN, all isolates were able to utilize KCN, although the latter was significantly more toxic; MICs ranged from 0.2 to 0.8 mM for KCN and greater than or equal to 50 mM for TCN. While growth occurred over a wide range of TCN concentrations (0.25 to 16 mM), degradation was most substantial under growth-limiting conditions and did not occur when ammonia was present. In addition, cells grown on TCN were found to accumulate nickel cyanide [Ni(CN)2] as a major biodegradation product. The results show that bacteria capable of growth on TCN can readily be isolated and that degradation (i) appears to parallel the capacity for growth on KCN, (ii) does not occur in the presence of ammonia, and (iii) proceeds via the formation of Ni(CN)2 as a biological metabolite.