Merck
  • Computational design, synthesis, and evaluation of miniproteins as androgen receptor coactivator mimics.
Computational design, synthesis, and evaluation of miniproteins as androgen receptor coactivator mimics.
Chemical communications (Cambridge, England) (2009-09-03)
Belen Vaz, Sabine Möcklinghoff, Simon Folkertsma, Scott Lusher, Jacob de Vlieg, Luc Brunsveld
ABSTRACT
Insertion of 3 to 4 mutations, based on in silico modelling, in a diverse set of natural miniproteins generates potent androgen receptor (AR) binders and a clear insight into the structure-activity relationship of such coactivator mimics concerning helix length.