Merck
Straightforward stereoselective access to cyclic peptidomimetics.
The Journal of organic chemistry (2009-05-15)
Santos Fustero, Natalia Mateu, Laia Albert, José Luis Aceña
ABSTRACT
The preparation of cyclic dipeptide mimetics from chiral imino lactones derived from (R)-phenylglycinol is described. Key steps of the synthetic route included the fully stereoselective construction of a quaternary center, the formation of six-, seven-, or eight-membered lactams by means of an RCM cyclization, and the introduction of a new amino group within the lactam ring. The synthesis of a tripeptide mimetic is also reported.
Sigma-Aldrich
N-(2-Hydroxyethyl)aniline, 98%