Merck
  • (R)-2,4-Dihydroxybutyramide seco-pseudonucleosides: new versatile homochiral synthons for synthesis of modified oligonucleotides.
(R)-2,4-Dihydroxybutyramide seco-pseudonucleosides: new versatile homochiral synthons for synthesis of modified oligonucleotides.
Organic letters (2002-12-20)
Natalia N Dioubankova, Andrey D Malakhov, Dmitry A Stetsenko, Vladimir A Korshun, Michael J Gait
ABSTRACT
[reaction: see text] Two series of seco-pseudonucleoside synthons were synthesized from (R)-(+)-alpha-hydroxy-gamma-butyrolactone and (R)-(-)-pantolactone by aminolysis, side-chain protection, dimethoxytritylation, and phosphitylation or solid-phase attachment. The phosphoramidites and solid supports were used in automated DNA synthesis to prepare oligonucleotides modified with one or more 2,4-dihydroxybutyramide units bearing side-chain reporter groups. These new oligonucleotide modification reagents allow the introduction of a label into any desired position within an oligonucleotide chain during solid-phase assembly.
Sigma-Aldrich
(R)-(+)-α-Hydroxy-γ-butyrolactone, 95%, optical purity ee: 98% (GLC)
Sigma-Aldrich
α-Hydroxy-γ-butyrolactone, technical grade
Sigma-Aldrich
Butyramide, ≥98.0% (T)