Merck
  • Antimicrobial peptides initiate IL-1 beta posttranslational processing: a novel role beyond innate immunity.
Antimicrobial peptides initiate IL-1 beta posttranslational processing: a novel role beyond innate immunity.
Journal of immunology (Baltimore, Md. : 1950) (2002-03-09)
David G Perregaux, Kanan Bhavsar, Len Contillo, Jishu Shi, Christopher A Gabel
ABSTRACT
Human monocytes stimulated with LPS produce large quantities of prointerleukin-1beta, but little of this cytokine product is released extracellularly as the mature biologically active species. To demonstrate efficient proteolytic cleavage and export, cytokine-producing cells require a secondary effector stimulus. In an attempt to identify agents that may serve as initiators of IL-1beta posttranslational processing in vivo, LPS-activated human monocytes were treated with several individual antimicrobial peptides. Two peptides derived from porcine neutrophils, protegrin (PTG)-1 and PTG-3, promoted rapid and efficient release of mature IL-1beta. The PTG-mediated response engaged a mechanism similar to that initiated by extracellular ATP acting via the P2X(7) receptor. Thus, both processes were disrupted by a caspase inhibitor, both were sensitive to ethacrynic acid and CP-424,174, two pharmacological agents that suppress posttranslational processing, and both were negated by elevation of extracellular potassium. Moreover, the PTGs, like ATP, promoted a dramatic change in monocyte morphology and a loss of membrane latency. The PTG response was concentration dependent and was influenced profoundly by components within the culture medium. In contrast, porcine neutrophil antimicrobial peptides PR-26 and PR-39 did not initiate IL-1beta posttranslational processing. The human defensin HNP-1 and the frog peptide magainin 1 elicited export of 17-kDa IL-1beta, but these agents were less efficient than PTGs. As a result of this ability to promote release of potent proinflammatory cytokines such as IL-1beta, select antimicrobial peptides may possess important immunomodulatory functions that extend beyond innate immunity.
Sigma-Aldrich
Magainin I, ≥97% (HPLC)
Sigma-Aldrich
CP-424,174, ≥98% (HPLC)