- Thymidine phosphorylase inhibitors with a homophthalimide skeleton.
Thymidine phosphorylase inhibitors with a homophthalimide skeleton.
Biological & pharmaceutical bulletin (2001-07-18)
T Kita, H Takahashi, Y Hashimoto
PMID11456133
ABSTRACT
Several N-phenylhomophthalimide derivatives were prepared and their inhibitory activity on thymidine phosphorylase/ platelet-derived endothelial cell growth factor (TP/PD-ECGF) was assessed. Among them, 2-(2,6-diethylphenyl)-7-nitro-1,2,3,4-tetrahydroisoquinoline-1,3-dione (9) was found to be a more potent inhibitor than the classical inhibitor, 5-nitrouracil (1). Lineweaver-Burk plot analysis indicated that 9 shows mixed-type competitive inhibition of TP/PD-ECGF, while 1 is a competitive inhibitor.