- A selective inversion recovery method for the improvement of 23Na NMR spectral resolution in isolated perfused rat hearts.
A selective inversion recovery method for the improvement of 23Na NMR spectral resolution in isolated perfused rat hearts.
NMR in biomedicine (1993-05-01)
T Simor, S K Kim, W J Chu, G M Pohost, G A Elgavish
PMID8347454
ABSTRACT
Shift-reagent-aided 23Na NMR spectroscopy allows differentiation of the intracellular (Na(i)) and extracellular sodium (Na(o)) signals. The goal of the present study has been to develop a 23Na NMR spectroscopic method to minimize the intensity of the shift-reagent-shifted Na(o) signal and thus increase Na(i) resolution. This is achieved by a selective inversion recovery (SIR) method which enhances the resolution between the Na(i) and Na(o) peaks in shift-reagent-aided 23Na NMR spectroscopy. The application of SIR with Dy(TTHA), Tm(DOTP), or with low concentrations of Dy(PPP)2 results in both good spectral resolution and physiologically acceptable contractile function in the isolated, perfused rat heart model.
Sigma-Aldrich
Triethylenetetramine-N,N,N′,N′′,N′′′,N′′′-hexaacetic acid, for complexometry, ≥98.0%