Merck
Faecal markers in metabolic balance studies.
Journal of human nutrition (1980-12-01)
S Bacon
ABSTRACT
The advantages and disadvantages of faecal markers that are available in the UK are discussed. Opinions and comments from units using them are included.
Sigma-Aldrich
Copper(I) thiocyanate, 99%