Merck
Binding of eugenol and o-ethoxybenzoic acid to bovine serum albumin.
Journal of dental research (1981-04-01)
S Fujisawa, E Masuhara
ABSTRACT
The binding constants (log K) of eugenol, o-ethoxy-benzoic acid, and their analogues, plus other organic compounds widely used in dentistry with bovine serum albumin, were examined with respect to the partition coefficients of these compounds in octanol-water (log P). Linear relationships between log K and log P values were obtained with high relative coefficients. Eugenol and o-ethoxybenzoic acid showed a relatively large log K value.
Sigma-Aldrich
2-Ethoxybenzoic acid, 98%