Merck
  • Roots and associated fungi drive long-term carbon sequestration in boreal forest.
Roots and associated fungi drive long-term carbon sequestration in boreal forest.
Science (New York, N.Y.) (2013-03-30)
K E Clemmensen, A Bahr, O Ovaskainen, A Dahlberg, A Ekblad, H Wallander, J Stenlid, R D Finlay, D A Wardle, B D Lindahl
ABSTRACT
Boreal forest soils function as a terrestrial net sink in the global carbon cycle. The prevailing dogma has focused on aboveground plant litter as a principal source of soil organic matter. Using (14)C bomb-carbon modeling, we show that 50 to 70% of stored carbon in a chronosequence of boreal forested islands derives from roots and root-associated microorganisms. Fungal biomarkers indicate impaired degradation and preservation of fungal residues in late successional forests. Furthermore, 454 pyrosequencing of molecular barcodes, in conjunction with stable isotope analyses, highlights root-associated fungi as important regulators of ecosystem carbon dynamics. Our results suggest an alternative mechanism for the accumulation of organic matter in boreal forests during succession in the long-term absence of disturbance.
Sigma-Aldrich
Ergosterol, ≥75%