Merck
  • Neurogliaform cells dynamically regulate somatosensory integration via synapse-specific modulation.
Neurogliaform cells dynamically regulate somatosensory integration via synapse-specific modulation.
Nature neuroscience (2012-12-12)
Ramesh Chittajallu, Kenneth A Pelkey, Chris J McBain
ABSTRACT
Despite the prevailing idea that neurogliaform cells produce a spatially unrestricted widespread inhibition, we demonstrate here that their activity attenuates thalamic-evoked feed-forward inhibition in layer IV barrel cortex but has no effect on feed-forward excitation. The result of this circuit selectivity is a dynamic regulation in the temporal window for integration of excitatory thalamic input, thus revealing a new role for neurogliaform cells in shaping sensory processing.
Sigma-Aldrich
CGP 55845 hydrochloride, ≥98% (HPLC)