Merck
  • Ursolic acid induces ER stress response to activate ASK1-JNK signaling and induce apoptosis in human bladder cancer T24 cells.
Ursolic acid induces ER stress response to activate ASK1-JNK signaling and induce apoptosis in human bladder cancer T24 cells.
Cellular signalling (2012-09-25)
Qing-you Zheng, Ping-ping Li, Feng-suo Jin, Chen Yao, Guo-hui Zhang, Tong Zang, Xing Ai
ABSTRACT
Here we studied the cellular mechanisms of ursolic acid's anti-bladder cancer ability by focusing on endoplasmic reticulum stress (ER stress) signaling. We show that ursolic acid induces a significant ER stress response in cultured human bladder cancer T24 cells. ER stress inhibitor salubrinal, or PERK silencing, diminishes ursolic acid-induced anti-T24 cell effects. Salubrinal inhibits ursolic acid-induced CHOP expression, Bim ER accumulation and caspase-3 activation in T24 cells. Ursolic acid induces IRE1-TRAF2-ASK1 signaling complex formation to activate pro-apoptotic ASK1-JNK signaling. We suggest that ER stress contributes to ursolic acid's effects against bladder cancer cells.
Product Description
Supelco
Ursolic acid, analytical standard
Sigma-Aldrich
Ursolic acid, ≥90%
Ursolic acid, primary reference standard