Merck
  • Palladium-catalyzed annulation of allenes with indole-2-carboxylic acid derivatives: synthesis of indolo[2,3-c]pyrane-1-ones via Ar-I reactivity or C-H functionalization.
Palladium-catalyzed annulation of allenes with indole-2-carboxylic acid derivatives: synthesis of indolo[2,3-c]pyrane-1-ones via Ar-I reactivity or C-H functionalization.
The Journal of organic chemistry (2012-07-26)
R Rama Suresh, K C Kumara Swamy
ABSTRACT
Two methodologies, one involving Ar-I reactivity and the other through C-H functionalization, for the formation of indolo[2,3-c]pyrane-1-ones via the corresponding allenes, are presented. A highly efficient approach to indolo[2,3-c]pyrane-1-one derivatives through the Pd-catalyzed regioselective annulation of allenes with 3-iodo-1-alkylindole-2-carboxylic acids is described. This method is fairly general for a wide range of allenes affording the respective indolo[2,3-c]pyrane-1-ones in good to excellent yields. In addition, a Pd(II)-catalyzed oxidative coupling of indole-2-caboxylic acid derivatives with allenes via direct C-H functionalization to afford the corresponding indolo[2,3-c]pyrane-1-ones in moderate to good yields has been developed.
Sigma-Aldrich
Indole-2-carboxylic acid, 98%