Merck
  • Development of a highly sensitive assay for enzyme-mediated reductive degradation of polychlorinated dibenzo-p-dioxin.
Development of a highly sensitive assay for enzyme-mediated reductive degradation of polychlorinated dibenzo-p-dioxin.
Environmental toxicology and chemistry (2012-03-27)
Yuzo Suzuki, Masaya Nakamura, Yuichiro Otsuka, Nao Suzuki, Keisuke Ohyama, Takeshi Kawakami, Kanna Sato, Shinya Kajita, Shojiro Hishiyama, Atsushi Takahashi, Yoshihiro Katayama
ABSTRACT
The degradation of 2-chloro-4,5-O-(4'-methyl-7', 8'-diphenyl)ether (CMDPE), an analog of 2,7-dichlorodibenzo-p-dioxin (2,7-DCDD), mediated by Geobacillus sp. UZO 3 cell-free extract was monitored. Ethyl acetate extracts of a complete reaction mixture incubated at 65°C for 18 h were analyzed either by thin layer chromatography (TLC) fractionation coupled with spectrometric detection or by gas chromatography-mass spectrometry (GC-MS). The reaction product 4-methylumbelliferone (4MU) was successfully isolated by TLC and visualized by a transilluminator at 450 nm. The 4MU, 4-chlorophenol, and reaction intermediate 6-chlorophenoxy-4-methylumbelliferone were all successfully detected by GC-MS. The presence of these compounds suggest that Geobacillus sp. UZO 3 cell-free extract also catalyzes the reductive cleavage of the diaryl ether bonds of CMDPE in a similar mechanism previously reported in 2,7-DCDD. In the present study, the authors describe a simple and highly sensitive fluorescent assay for a new dioxin degrading enzyme(s).
Sigma-Aldrich
4-Chlorophenol, ≥99%