Merck
  • Processing ulvan into 2D structures: cross-linked ulvan membranes as new biomaterials for drug delivery applications.
Processing ulvan into 2D structures: cross-linked ulvan membranes as new biomaterials for drug delivery applications.
International journal of pharmaceutics (2012-01-28)
Anabela Alves, Elisabete D Pinho, Nuno M Neves, Rui A Sousa, Rui L Reis
ABSTRACT
The polysaccharide ulvan, composed of sulphated rhamnose, glucoronic and iduronic acids was used to produce polymeric membranes by solvent casting. As ulvan is soluble in water, a cross-linking step was necessary to render the membrane insoluble in water and stable at physiological conditions. Cross-linked ulvan membranes were characterized by FTIR, SEM, swelling behaviour was investigated and the mechanical performance assessed by quasi-static tensile testing. Furthermore, the ability and mechanism of sustained release of a model drug from ulvan membranes was investigated. Produced membranes revealed remarkable ability to uptake water (up to ∼1800% of its initial dry weight) and increased mechanical performance (1.76 MPa) related with cross-linking. On the other hand, medicated ulvan dressings demonstrate the potential as drug delivery devices. Using a model drug we have observed an initial steady release of the drug - of nearly 49% - followed by slower and sustained release up to 14 days. The properties of ulvan membranes herein revealed suggest a great potential of this natural sulphated polysaccharide as a wound dressing.