Merck
  • Benzocaine-loaded polymeric nanocapsules: study of the anesthetic activities.
Benzocaine-loaded polymeric nanocapsules: study of the anesthetic activities.
Journal of pharmaceutical sciences (2011-11-23)
Nathalie Ferreira Silva De Melo, Daniele Ribeiro De Araújo, Renato Grillo, Carolina Morales Moraes, Angélica Prado De Matos, Eneida de Paula, André Henrique Rosa, Leonardo Fernandes Fraceto
ABSTRACT
This paper describes a comparison of different polymeric nanocapsules (NCs) prepared with the polymers poly(D,L-lactide-co-glycolide), poly(L-lactide) (PLA), and poly(ε-caprolactone) and used as carrier systems for the local anesthetic (LA) benzocaine (BZC). The systems were characterized and their anesthetic activities investigated. The results showed particle size distributions with polydispersity indices below 0.135, average diameters up to 120 nm, zeta potentials up to -30 mV, and entrapment efficiencies around 70%. Formulations of BZC using the polymeric NCs presented slower release profiles, compared with that of free BZC. Slowest release (release constant, k = 0.0016 min(-1)) was obtained using the PLA NC system. Pharmacological evaluation showed that encapsulation of BZC in PLA NCs prolonged its anesthetic action. This new formulation could potentially be used in future applications involving the gradual release of local anesthetics (LAs).
Product Description
Supelco
Benzocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Benzocaine, ≥99% (HPLC)
Sigma-Aldrich
Ethyl 4-aminobenzoate, 98%