Merck
  • Investigation into the regiochemistry of some isoxazoles derived from 1,3-dipolar cycloaddition of 4-nitrobenzonitrile oxide with some dipolarophiles: a combined theoretical and experimental studies.
Investigation into the regiochemistry of some isoxazoles derived from 1,3-dipolar cycloaddition of 4-nitrobenzonitrile oxide with some dipolarophiles: a combined theoretical and experimental studies.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-06-07)
N Dorostkar-Ahmadi, M Bakavoli, F Moeinpour, A Davoodnia
ABSTRACT
Reaction of 4-nitrobenzonitrile oxide (2) which was generated in situ with acrylo nitrile (3), methyl methacrylate (4) and allyl bromide (5) as dipolarphile afforded the new 7a, 8a and 9a compounds respectively. Reactivity and regiochemistry of these reactions were investigated using activation energy calculations and density functional theory (DFT)-based reactivity indexes. The theoretical 13C NMR chemical shifts of the cycloadducts which were obtained by GIAO method were comparable with the observed values.
Sigma-Aldrich
4-Nitrobenzonitrile, 97%