Merck
  • Competitive interaction of three peroxidizing herbicides with the binding of [3H]acifluorfen to corn etioplast membranes.
Competitive interaction of three peroxidizing herbicides with the binding of [3H]acifluorfen to corn etioplast membranes.
FEBS letters (1990-10-15)
R Varsano, M Matringe, N Magnin, R Mornet, R Scalla
ABSTRACT
The specific binding of the herbicide acifluorfen 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid to corn etioplast membranes is competitively inhibited by protoporphyrinogen IX, the substrate of protoporphyrinogen oxidase. Three other peroxidizing molecules, oxadiazon [5-terbutyl-3-(2,4-dichloro-5-isopropoxyphenyl)-1,3,4-oxadiazol -2-one], LS 82556 [(S)3-N-(methylbenzyl)carbamoyl-5-propionyl-2,6-lutidine], and M&B 39279 [5-amino-4-cyano-1-(2,6-dichloro-4-trifluoromethylphenyl)pyrazol], also compete with acifluorfen for its binding site. The four herbicides thus bind to the same site, or to closely located sites, on the enzyme protoporphyrinogen oxidase.
Product Description
Supelco
Oxadiazon, PESTANAL®, analytical standard