Merck
  • Supramolecular control of fluorescence through reversible encapsulation.
Supramolecular control of fluorescence through reversible encapsulation.
Journal of the American Chemical Society (2010-07-09)
Henry Dube, Mark R Ams, Julius Rebek
ABSTRACT
The trans to cis isomerization of 4,4'-dimethylazobenzene by the nonchemical stimuli light and heat alters its guest binding properties, allowing control over encapsulation of a second guest. We show here how this remote control for reversible encapsulation can be used as a supramolecular fluorescence on/off switch. If trans-4-ethyl-4'-methylstilbene is used as the second guest its fluorescence is altered depending on whether it is free in solution or encapsulated. We demonstrate that the change in fluorescence is indeed a consequence of the controlled encapsulation state of the stilbene by correlation of fluorescence and (1)H NMR data.
Sigma-Aldrich
Methylstyrene, 60% meta, 40% para and 1% ortho, 99%, contains ~50 ppm 4-tert-butylcatechol as inhibitor