Merck
Determination of sialic acids in milks and milk-based products.
Analytical biochemistry (2010-06-18)
Véronique Spichtig, Julien Michaud, Sean Austin
ABSTRACT
Sialic acids are becoming recognized as important components of milk-based products for infants and young children. As such, many companies now label the sialic acid content of their products. To control the labeling, suitable methods are required for this analysis. The objective of this work was to set up a rapid and sensitive method for the determination of the two most commonly occurring sialic acids, N-acetylneuraminic acid (Neu5Ac) and N-glycolylneuraminic acid (Neu5Gc), using high-performance liquid chromatography (HPLC). The sialic acids were released from their parent oligosaccharides, glycoproteins, or glycolipids by mild acid hydrolysis using formic acid. They were then derivatized using 1,2-diamino-4,5-methylenedioxybenzene (DMB) and subsequently separated on a Zorbax SB-Aq Rapid Resolution column in less than 2 min. The method developed was validated on various milk-based products and ingredients containing sialic acid at levels from 0.3 to 900 mg/100 g. Spiking experiments indicate that the sialic acid recoveries ranged from 87% to 108%. The expanded measurement uncertainty was typically below 15% for Neu5Gc and typically below 10% for Neu5Ac or the sum of the sialic acids, with a few exceptions. The proposed method is fast, specific, and easy to set up for compliance analysis in a routine laboratory.
Sigma-Aldrich
N-Glycolylneuraminic acid, ≥95% (HPLC), semisynthetic