Merck
  • Chiral N-phosphonyl imine chemistry: an efficient asymmetric synthesis of chiral N-phosphonyl propargylamines.
Chiral N-phosphonyl imine chemistry: an efficient asymmetric synthesis of chiral N-phosphonyl propargylamines.
Organic & biomolecular chemistry (2010-02-19)
Parminder Kaur, Gaurav Shakya, Hao Sun, Yi Pan, Guigen Li
ABSTRACT
A variety of substituted chiral propargylamines have been synthesized by reacting chiral N-phosphonylimines with lithium aryl/alkyl acetylides. Seventeen examples were studied to give excellent yields (>90%) and diastereoselectivities (96 : 4 to 99 : 1). It was found that the types of bases for generating acetylides and solvents are crucial for effectiveness of this asymmetric reaction. In addition, N,N-isopropyl group on chiral N-phosphonylimine auxiliary was proven to be superior to other protecting groups in controlling diastereoselectivity.
Sigma-Aldrich
Propargylamine, 98%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€70.90
25 g
Please contact Customer Service for Availability
€178.00
Sigma-Aldrich
Propargylamine hydrochloride, 95%
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
5 g
Please contact Customer Service for Availability
€70.90
25 g
Please contact Customer Service for Availability
€178.00