Merck
Supramolecular rhombic grids formed from bimolecular building blocks.
Journal of the American Chemical Society (2009-09-03)
Neng-Fang She, Meng Gao, Xiang-Gao Meng, Guang-Fu Yang, Johannes A A W Elemans, An-Xin Wu, Lyle Isaacs
ABSTRACT
A programmed assembly process produces rhombic grid networks from compounds L1-L4 in the crystal by pi-pi stacking interactions that generate a bimolecular grid synthon, which undergoes further NH...N hydrogen-bond-mediated assembly.
Sigma-Aldrich
Glycoluril, 97%