Merck
  • Enantioselective monoterpene alcohol acetylation in Origanum, Mentha and Salvia species.
Enantioselective monoterpene alcohol acetylation in Origanum, Mentha and Salvia species.
Phytochemistry (2008-10-07)
Olga Larkov, Alon Zaks, Einat Bar, Efraim Lewinsohn, Nativ Dudai, Alfred M Mayer, Uzi Ravid
ABSTRACT
Selected plants within the Origanum, Mentha and Salvia genera, that contain significant amounts of chiral volatile alcohols and their related acetates, exhibit remarkable enantioselectivity of alcohol acetyl transferase (AAT) activity and particularly can discriminate between linalool enantiomers. Origanum dayi AAT produced almost enantiomerically pure (R)-linalyl acetate by enzymatic acetylation of racemic linalool, whereas the closely related O. majorana AAT produced a mixture of (R)- and (S)-linalyl acetate with a ratio of 6:4. V(max) of O. dayi acetylation activity was 30-fold higher for (R)-linalool, whereas in O. majorana no such differences were found.
Sigma-Aldrich
Linalyl acetate, ≥97%, FCC, FG
Sigma-Aldrich
Linalyl acetate, natural, ≥96%, FG
Sigma-Aldrich
3,7-Dimethyl-1,6-octadien-3-yl acetate, 97%