Merck
  • Influence of threose nucleoside units on the catalytic activity of a hammerhead ribozyme.
Influence of threose nucleoside units on the catalytic activity of a hammerhead ribozyme.
Chemistry & biodiversity (2006-12-29)
Veerle Kempeneers, Mattheus Froeyen, Karen Vastmans, Piet Herdewijn
ABSTRACT
TNA (alpha-L-threose nucleic acids) is potentially a natural nucleic acid, that might have acted as an evolutionary alternative of RNA. We determined the catalytic activity of hammerhead ribozymes containing a threofuranosyl-modified nucleoside at position U4 and U7, and compared these results with those obtained from HNA (hexitol nucleic acids) insertion into the same ribozyme. Our experiments showed that, although the threofuranosyl-modified ribozymes still cleave the substrate strand, cleavage activity is highly decreased. It, therefore, seems that TNA can play a functional role in the RNA world, but only to a limited extent.