Skip to Content
Fechas de fin de año: Facturación de pedidos para envío en Ciudad de México, zona Metropolitana del Valle de México y Toluca hasta 13/12. Para otras regiones, hasta el 11/12. Receso de Merck Mexico del 18/12 al 02/01.
Merck
Adsorption of Chrysoidine R by using fly ash in batch process.
Journal of hazardous materials (2006-12-05)
Manickam Matheswaran, Thirugnanam Karunanithi
ABSTRACT
This investigation deals with effective utilization of fly ash as adsorbent for the removal of Chrysoidine R from the aqueous solution. The fly ash is a major byproduct generated in coal-based thermal power plants and has good potential for use as an adsorbent. A series of experiments were carried out in a batch adsorption technique to obtain the effect of process variables viz. contact time, pH (2, 4, 6 and 8) initial concentration of the dye (400, 600, 800 and 1000mgL(-1)), amount of the adsorbent (125, 250, 375 and 500mgL(-1)), and temperature (303, 313, 323 and 333K) on adsorption. The concentration of dye was determined by spectrophotometer. The results showed that as the amount of the adsorbent was increased, the percentage of dye removal increased accordingly; higher adsorption percentage was observed at lower concentration of chrysoidine. The adsorption data were analyzed using Langmuir and Freundlich isotherms. The adsorption was found to obey pseudo-first order kinetics. An intra particle diffusion model was used to fit the experimental data. The thermodynamic parameters such as standard change in free energy, enthalpy and entropy of adsorption have been calculated. Adsorption of Chrysoidine R on fly ash was found to be an exothermic reaction.
Sigma-Aldrich
Chrysoidine G, for microscopy (Bact., Bot., Vit.)
Pricing and availability is not currently available. Contact Technical Service
Supelco
Chrysoidine G, analytical standard
Pricing and availability is not currently available. Contact Technical Service